missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myeloid leukemia factor 1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93971-0.02ml
This item is not returnable.
View return policy
Description
Myeloid leukemia factor 1 Polyclonal antibody specifically detects Myeloid leukemia factor 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Myeloid leukemia factor 1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Myelodysplasia-myeloid leukemia factor 1, myeloid leukemia factor 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-268 of human MLF1 (NP_071888.1). MFRMLNSSFEDDPFFSESILAHRENMRQMIRSFSEPFGRDLLSISDGRGRAHNRRGHNDGEDSLTHTDVSSFQTMDQMVSNMRNYMQKLERNFGQLSVDPNGHSFCSSSVMTYSKIGDEPPKVFQASTQTRRAPGGIKETRKAMRDSDSGLEKMAIGHHIHDRAHVIKKSKNKKTGDEEVNQEFINMNESDAHAFDEEWQSEVLKYKPGRHNLGNTRMRSVGHENPGSRELKRREKPQQSPAIEHGRRSNVLGDKLHIKGSSVKSNKK | |
| 0.02 mL | |
| Cancer, Stem Cell Markers | |
| 4291 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction