Learn More
Myeloperoxidase/MPO Antibody (CL14991) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26846JF669
Product Code. 30291980
  • 737.00 € / 0.10mL
Description
Myeloperoxidase/MPO Monoclonal antibody specifically detects Myeloperoxidase/MPO in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Myeloperoxidase/MPO
Monoclonal
Janelia Fluor 669
EC 1.11.1, EC 1.11.1.7, myeloperoxidase
This antibody was developed using a synthetic peptide derived from P05164, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism
4353
Store at 4⪚C in the dark.
IgG3
Immunohistochemistry, Western Blot
CL14991
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified