missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myeloperoxidase/MPO Antibody (rMPO/8694), Novus Biologicals™
Mouse Monoclonal Antibody
249.00 € - 494.00 €
Specifications
| Antigen | Myeloperoxidase/MPO |
|---|---|
| Clone | rMPO/8694 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18632574
|
Novus Biologicals
NBP3-20650-100ug |
100 μg |
494.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18662794
|
Novus Biologicals
NBP3-20650-20ug |
20 μg |
249.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Myeloperoxidase/MPO Monoclonal antibody specifically detects Myeloperoxidase/MPO in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Myeloperoxidase/MPO | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Immunology, Innate Immunity, Lipid and Metabolism | |
| 10mM PBS with 0.05% BSA | |
| 4353 | |
| IgG1 κ | |
| Protein A or G purified |
| rMPO/8694 | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 1.11.1, EC 1.11.1.7, myeloperoxidase | |
| Recombinant fragment (around aa150-250) of human Myeloperoxidase/MPO protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title