missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Myeloperoxidase Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA516672

Product Code. 11335833

  • 506.00 € / 500µL

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Description

Description

PA5-16672 targets Myeloperoxidase in IHC (P) applications and shows reactivity with Human, mouse, and Rat samples. The PA5-16672 immunogen is purified human granulocytic MPO. PA5-16672 detects MPO-DNA complexes in purified PMA-induced human neutrophil extracellular traps in ELISA application. Concentration is not determined for this particular antibody. The final product is manufactured from stock material based upon a dilution ratio. The antibody is still quality tested to ensure expected performance and lot-to-lot consistency.

Myeloperoxidase (MPO) is a heme protein that is synthesized during myeloid differentiation and is a major component of neutrophil azurophilic granules. It is produced as a single chain precursor and subsequently cleaved into a light and heavy chain. The mature MPO is a tetramer composed of 2 light chains and 2 heavy chains. This enzyme is responsible for producing hypohalous acids that are central to the microbicidal activity of neutrophils. Diseases that are associated with MPO include Myeloperoxidase Deficiency and Alzheimer's Disease, Familial, 1.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Myeloperoxidase
Polyclonal
Unconjugated
MPO
84 kDa myeloperoxidase; 89 kDa myeloperoxidase; mKIAA4033; MPO; Myeloperoxidase; Myeloperoxidase heavy chain; Myeloperoxidase light chain
Rabbit
Purified
RUO
17523, 303413, 4353
4°C
Liquid
ELISA, Immunohistochemistry (Paraffin)
Conc. Not Determined
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
P05164, P11247
MPO
Purified human granulocytic MPO.
500 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.