missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYF5 Recombinant Rabbit Monoclonal Antibody (24GB615)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557092
This item is not returnable.
View return policy
Description
MYF5 Recombinant Monoclonal Antibody for Western Blot, Flow
Myogenic factor 5 (MYF5) is a gene located on chromosome 12q21.31, encoding a basic helix-loop-helix (bHLH) transcription factor essential for skeletal muscle development and differentiation. MYF5 is one of the key members of the myogenic regulatory factor (MRF) family, which also includes MyoD, myogenin, and MRF4. It plays a crucial role in myogenesis by regulating the expression of genes necessary for muscle precursor cell proliferation and differentiation into mature muscle fibers. During embryonic development, MYF5 is involved in the formation of muscle tissue and maintenance of muscle progenitor cells. Mutations or dysregulation in MYF5 can lead to developmental abnormalities in muscle formation and are implicated in muscle-related disorders. Research into MYF5 continues to explore its regulatory mechanisms in muscle biology and potential therapeutic applications in muscular dystrophies and muscle repair processes.
Specifications
| MYF5 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| MYF5 | |
| B130010J22Rik; basic helix-loop-helix nuclear protein; bHLHc2; cb641; Class C basic helix-loop-helix protein 2; muscle regulatory protein Myf5; MYF5; myf-5; myf5 protein; MyoD family; myogenic factor 5; myogenic factor-5; myogenic regulatory factor 5; myogenic regulatory factor MYF-5; unnamed protein product; zgc:110509 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4617 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB615 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P13349 | |
| MYF5 | |
| A synthesized peptide derived from human MYF5 (aa 201-255). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction