Learn More
Snail Antibody (20C8), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50300AF647
Product Code. 18108038
  • 464.00 € / 0.10mL
Description
Snail Monoclonal specifically detects Snail in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Snail
Monoclonal
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
SNAI1
Human Snail recombinant protein
0.1 mL
Breast Cancer, Cancer, Developmental Biology, Hypoxia, Lipid and Metabolism, Metastasis, Transcription Factors and Regulators
6615
Store at 4C in the dark.
IgG2a κ
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
20C8
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Protein sna, Protein snail homolog 1, SLUGH2, SNA, SNAHdJ710H13.1, snail 1 (drosophila homolog), zinc finger protein, snail 1 homolog, snail 1 zinc finger protein, snail 1, zinc finger protein, snail homolog 1 (Drosophila), zinc finger protein SNAI1
Mouse
Protein A purified
RUO
Primary
Human
Purified