Learn More
MYH10-Specific Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-19673-100UL
 View more versions of this product
Product Code. 18089685
  • 450.00 € / 100µL
Description
The MYH10-Specific antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human MYH10-Specific. This antibody recognizes human, mouse, rat antigen. The MYH10-Specific antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
MYH10-Specific
1000 μg/mL
MYH10, Myosin 10, Myosin heavy chain 10
100 μL
4628
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat
150 kDa