missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYH7B Antibody (SR1337), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22346-50ul
This item is not returnable.
View return policy
Description
MYH7B Monoclonal antibody specifically detects MYH7B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| MYH7B | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 57644 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| SR1337 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| Antigen MLAA-21, MHC14, MYH14, Myosin cardiac muscle beta chain, myosin heavy chain 7B, Myosin heavy chain 7B, cardiac muscle beta isoform, Myosin-7B, Slow A MYH14 | |
| A synthesized peptide derived from human MYH7B (Uniprot #: A7E2Y1) | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction