missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYH8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572846
This item is not returnable.
View return policy
Description
MYH8 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Myosin is a highly conserved, ubiquitously expressed protein that interacts with Actin to generate the force for cellular movements. Conventional Myosins are hexameric proteins consisting of two heavy chain subunits, a pair of non-phosphorylatable light chain subunits and a pair of phosphorylatable light chain subunits. Three general classes of Myosin have been cloned: smooth muscle Myosins, striated muscle Myosins and non-muscle Myosins. Contractile activity in smooth muscle is regulated by the calcium/calmodulin-dependent phosphorylation of Myosin light chain (MLC) by Myosin light chain kinase. Myosin heavy chains, which are encoded by the MYH gene family, contain Actin-activated ATPase activity which generates the motor function of Myosin. Myosin heavy chains were initially isolated from a human fetal skeletal muscle and are the major determinant in the speed of contraction of skeletal muscle. Various isoforms of myosin heavy chains are differentially expressed depending on the functional activity of the muscle.
Specifications
| MYH8 | |
| Polyclonal | |
| Unconjugated | |
| MYH8 | |
| 4832426G23Rik; AI327267; Beta-MHC antisense; DA7; fetal-myosin heavy chain; gtMHC-F; MHCp; Myh8; MyHC-peri; MyHC-perinatal; MyHC-pn; Myhsp; Myhs-p; Myosin 8; Myosin heavy chain 8; myosin heavy chain, skeletal muscle, perinatal; myosin, heavy chain 8, skeletal muscle, perinatal; myosin, heavy polypeptide 8, skeletal muscle, perinatal; Myosin8; myosin-8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17885, 252942, 4626 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P04462, P13535, P13542 | |
| MYH8 | |
| MYH8 antibody for a 19 amino acid peptide near the amino terminus of human MYH8. The immunogen is within amino acids 60 - 110 of MYH8. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction