missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MYH9 Recombinant Rabbit Monoclonal Antibody (K01_2N59)
GREENER_CHOICE

Product Code. 30825487
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30825487 100 μL
30830360 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30825487 Supplier Invitrogen™ Supplier No. MA564143

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

MYH9 Recombinant Monoclonal Antibody for IHC (P), Western Blot

The MYH9 gene, located on chromosome 22q12.3, encodes the heavy chain of non-muscle myosin IIA (NMHC IIA), a critical component of the actin cytoskeleton that plays essential roles in various cellular processes. Structurally, the MYH9 gene spans over 106 kilobases and includes 41 exons that translate into a protein of 1,960 amino acids. This protein is a part of a hexameric complex, which includes two heavy chains, two regulatory light chains, and two essential light chains. The NMHC IIA protein interacts with actin filaments and is involved in cellular activities such as cell migration, adhesion, division, and maintenance of cell shape. Functionally, mutations in MYH9 can result in a spectrum of autosomal dominant disorders collectively known as MYH9-related diseases (MYH9-RD), which include conditions such as May-Hegglin anomaly, Fechtner syndrome, and Epstein syndrome. These disorders are primarily characterized by macrothrombocytopenia (abnormally large platelets) and may lead to other complications such as hearing loss, renal failure, and cataracts later in life. MYH9 is also crucial in hematopoiesis, where its proper function is necessary for the survival and maintenance of hematopoietic stem and progenitor cells (HSPCs). Loss of MYH9 function disrupts normal hematopoiesis, leading to severe blood cell deficiencies and bone marrow failure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYH9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_2N59
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene MYH9
Gene Accession No. P35579, Q62812
Gene Alias BDPLT6; Cellular myosin heavy chain, type A; DFNA17; epsts; fi22c04; fj85e11; flectin; Fltn; FTNS; KLG/PTK7; LOC100911597; LOW QUALITY PROTEIN: myosin-9; MGC104539; MHA; Myh9; myh9.L; myh9a; myh9l2; Myhn1; Myhn-1; myosin; Myosin Heavy Chain 2A; myosin heavy chain 9; myosin heavy chain IX; myosin heavy chain, non-muscle IIa; myosin IIA; myosin, heavy chain 9, non-muscle; myosin, heavy chain 9, non-muscle L homeolog; myosin, heavy chain 9, non-muscle, like-2; myosin, heavy chain 9a, non-muscle; myosin, heavy polypeptide 9, non-muscle; myosin, heavy polypeptide 9a, non-muscle; myosin-2a; myosin9; Myosin-9; myosin-9; myosin-9a; myosin-9-like; NMHC II-A; NMHC-II A; NMHCIIA; NMHC-II-A; NMMHC IIA; NMMHC II-a; nmmhca; NMMHC-A; NMMHC-IIA; non-muscle myosin heavy chain 9; non-muscle myosin heavy chain A; non-muscle myosin heavy chain II A; non-muscle myosin heavy chain IIa; nonmuscle myosin heavy chain II-A; non-muscle myosin heavy polypeptide 9; nonmuscle myosin II heavy chain A; TU72.6; Unknown (protein for IMAGE:7151033); Unknown (protein for IMAGE:7177375); Unknown (protein for IMAGE:7268224); wu:fi22c04; wu:fi22c04 protein; wu:fj85e11; XELAEV_18023533mg; zgc:162029; zgc:66164; zNMHC-IIA
Gene Symbols MYH9
Host Species Rabbit
Immunogen A synthetic peptide of human non-muscle Myosin IIA.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100911597, 4627
Target Species Human, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.