missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYL6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00 € - 624.00 €
Specifications
| Antigen | MYL6 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18444312
|
Novus Biologicals
NBP2-30441-25ul |
25ul |
415.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18188672
|
Novus Biologicals
NBP2-30441 |
0.1 mL |
624.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MYL6 Polyclonal specifically detects MYL6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MYL6 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| P60660 | |
| 4637 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: MTEEEVEMLVAGHEDSNGCINYEAF | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Polyclonal | |
| Rabbit | |
| Vision | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ESMLC, LC17, LC17A, LC17B, LC17-GI, LC17-NM, MLC1SM, MLC-3, MLC3NM, MLC3SM, Myosin light chain 3, Myosin light chain A3, Myosin light chain alkali 3, myosin light polypeptide 6,17 kDa myosin light chain, myosin, light chain 6, alkali, smooth muscle and non-muscle, myosin, light polypeptide 6, alkali, smooth muscle and non-muscle, Smooth muscle and nonmuscle myosin light chain alkali 6 | |
| MYL6 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts