missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MYO1E Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554913
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Immunogen sequence: VIRNQYVPYP HAPGSQRSNQ KSLYTSMARP PLPRQQSTSS DRVSQTPESL DFLKVPDQGA AGVRRQTTSR PPPAGGRPKP QPKPKPQVPQ CKALYAYDAQ DTDELSFNAN DIIDIIKEDP SGWWTGRLRG KQG Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 92%.
Myosins are actin-based motor molecules with ATPase activity. Unconventional myosins serve in intracellular movements. Their highly divergent tails are presumed to bind to membranous compartments, which would be moved relative to actin filaments.
Specifikationer
| MYO1E | |
| Polyclonal | |
| Unconjugated | |
| MYO1E | |
| 2310020N23Rik; 9130023P14Rik; AA407778; FSGS6; HuncM-IC; LOW QUALITY PROTEIN: unconventional myosin-Ie; molecular motor; MYO1C; Myo1e; MYO1E variant protein; myosin heavy chain myr 3; myosin IE; myosin-1e; myosin-IC; myosin-Ie; myosin-Ie; unconventional myosin-Ie; myr 3; Myr3; MYR5; unconventional myosin 1E; Unconventional myosin from rat 5; unconventional myosin-Ie | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4643 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q12965 | |
| MYO1E | |
| Recombinant protein corresponding to Human MYO1E. Recombinant protein control fragment (Product #RP-101032). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering