missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myoferlin Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94759-0.1ml
This item is not returnable.
View return policy
Description
Myoferlin Polyclonal antibody specifically detects Myoferlin in Human, Mouse samples. It is validated for Western Blot
Specifications
| Myoferlin | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| fer-1 (C.elegans)-like 3 (myoferlin), FER1L3, fer-1-like 3, myoferlin, fer-1-like 3, myoferlin (C. elegans), Fer-1-like protein 3, FLJ36571, FLJ90777, KIAA1207, myoferlin | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human MYOF (NP_038479.1). MLRVIVESASNIPKTKFGKPDPIVSVIFKDEKKKTKKVDNELNPVWNEILEFDLRGIPLDFSSSLGIIVKDFETIGQNKLIGTATVALKDLTGDQSRSLPYKLISLLNEKGQDTGATIDLVIGYDPPSAPHPNDLSGPSVPGMGGDGEEDEGDEDRLDNAVRGPGPKGPVGTVSEAQLARRLTKVKNSRR | |
| 0.1 mL | |
| Cardiovascular Biology, Cell Biology | |
| 26509 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction