Learn More
Myosin heavy chain 11 Antibody (CL14619) [DyLight 550], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26875R
Product Code. 30302726
  • 770.00 € / 0.10mL
Description
Myosin heavy chain 11 Monoclonal antibody specifically detects Myosin heavy chain 11 in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Myosin heavy chain 11
Monoclonal
DyLight 550
AAT4, FAA4, MYH11, MYH11 myosin, heavy chain 11, smooth muscle, SMHC, SMMHC, Smooth muscle myosin heavy chain
This antibody was developed using a recombinant protein derived from P35749, with the exact immunogen sequence remaining proprietary.
0.1 mL
Hypoxia, Stem Cell Markers
4629
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Western Blot
CL14619
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified