Learn More
myosin X Antibody (CL9025) [DyLight 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-88927G
Product Code. 30307357
  • 737.00 € / 0.10mL
Description
myosin X Monoclonal antibody specifically detects myosin X in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
myosin X
Monoclonal
DyLight 488
FLJ10639, FLJ21066, FLJ22268, KIAA0799FLJ43256, MGC131988, myosin X, myosin-X, Unconventional myosin-10
This antibody was developed using a synthetic peptide derived from Q9HD67, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cell Biology, Cytoskeleton Markers, Signal Transduction
4651
Store at 4⪚C in the dark.
IgG1
Western Blot
CL9025
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified