missing translation for 'onlineSavingsMsg'
Learn More

Myricetin, 98%

Product Code. 15443219
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15443219 50 mg 50mg
15423219 10 mg 10mg
15433219 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 15443219 Lieferant Thermo Scientific Alfa Aesar Lieferanten-Nr. J60450.MF
 Restricted Item
In Stock
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 529-44-2 | C15H10O8 | 318.24 g/mol

Myricetin is used as an antioxidant. It is a cell-permeable flavonoid used in inflammatory, diabetes, and cancer studies. It is effective in protecting neurons against oxidative stresses. Further, it inhibits yeast alfa-glucosidase,1 glyoxalase I in vitro and bovine milk xanthine oxidase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Myricetin is used as an antioxidant. It is a cell-permeable flavonoid used in inflammatory, diabetes, and cancer studies. It is effective in protecting neurons against oxidative stresses. Further, it inhibits yeast alfa-glucosidase,1 glyoxalase I in vitro and bovine milk xanthine oxidase.

Solubility
Soluble in dimethyl sulfoxide, dimethyl formamide and ethanol. Insoluble in water.  

Notes
Store in a cool place. Incompatible with strong oxidizing agents.    
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 529-44-2
Molecular Formula C15H10O8
Molecular Weight (g/mol) 318.24
MDL Number MFCD00006827
InChI Key IKMDFBPHZNJCSN-UHFFFAOYSA-N
Synonym myricetin, cannabiscetin, myricetol, myricitin, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl-4h-chromen-4-one, 3,3',4',5,5',7-hexahydroxyflavone, 3,5,7,3',4',5'-hexahydroxyflavone, unii-76xc01ftoj, ccris 5838, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl-4h-1-benzopyran-4-one
PubChem CID 5281672
ChEBI CHEBI:18152
SMILES OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC(O)=C(O)C(O)=C2)=C1

Spezifikation

Melting Point >300°C
Color Brown-Yellow
Quantity 50 mg
Beilstein 332331
Merck Index 14,6332
Solubility Information Soluble in dimethyl sulfoxide, dimethyl formamide and ethanol; Insoluble in water .
IUPAC Name 3,5,7-trihydroxy-2-(3,4,5-trihydroxyphenyl)chromen-4-one
Formula Weight 318.24
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material Myricetin
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.