missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Naphthyl)-N-(phenylmethylene)amine, 97%, Thermo Scientific™

Product Code. 10260153
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10260153 1 g Amber Glass Bottle 1g
10567493 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10260153 Supplier Thermo Scientific Maybridge Supplier No. AC33318DA
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 890-51-7
Molecular Formula C17H13N
Molecular Weight (g/mol) 231.298
MDL Number MFCD02235774
InChI Key NQPYSYDZNCHIQY-UHFFFAOYSA-N
Synonym n-1-naphthyl-n-phenylmethylene amine, n-benzylidenenaphthalen-1-amine, n-e-phenylmethylidene naphthalen-1-amine, 1z-1-naphthyl-2-phenyl-1-azaethene, n-naphthalen-1-yl-1-phenylmethanimine, 1z-n-naphthalen-1-yl-1-phenylmethanimine, n-benzylidene-1-napthylamine, benzylidene-a-napthylamine, maybridge3_000265, n-benzylidene-1-naphthalenamine
PubChem CID 1201463
IUPAC Name N-naphthalen-1-yl-1-phenylmethanimine
SMILES C1=CC=C(C=C1)C=NC2=CC=CC3=CC=CC=C32

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 231.3
Chemical Name or Material N-(1-naphthyl)-N-(phenylmethylene)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.