missing translation for 'onlineSavingsMsg'
Learn More

N-(2,4-Dichlorophenyl)urea, TRC

Product Code. 30134542
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134542 1 g 1g
30009749 250 mg 250mg
30004338 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134542 Supplier Toronto Research Chemicals Supplier No. TRCD4375731G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5428-50-2
Molecular Formula C7 H6 Cl2 N2 O
Molecular Weight (g/mol) 205.04
Synonym N-(2,4-Dichlorophenyl)urea, (2,4-Dichlorophenyl)urea, 1-(2,4-Dichlorophenyl)urea;
IUPAC Name (2,4-dichlorophenyl)urea
SMILES NC(=O)Nc1ccc(Cl)cc1Cl

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H6Cl2N2O/c8-4-1-2-6(5(9)3-4)11-7(10)12/h1-3H,(H3,10,11,12)
Formula Weight 203.9857
Chemical Name or Material 1-(2,4-Dichlorophenyl)urea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.