missing translation for 'onlineSavingsMsg'
Learn More

N-(2,6-Dimethylphenyl)acetamide, TRC

Product Code. 30138725
Change view
Click to view available options
Quantity:
100 mg
1 g
50 g
Unit Size:
100mg
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138725 50 g 50g
30003328 1 g 1g
30011197 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138725 Supplier Toronto Research Chemicals Supplier No. TRCD44668050G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2198-53-0
Molecular Formula C10 H13 N O
Molecular Weight (g/mol) 163.22
Synonym N-(2,6-Dimethylphenyl)-acetamide, Lidocaine Hydrochloride Imp. C (EP)
IUPAC Name N-(2,6-dimethylphenyl)acetamide
SMILES CC(=O)Nc1c(C)cccc1C

Specifications

Quantity 50 g
InChI Formula InChI=1S/C10H13NO/c1-7-5-4-6-8(2)10(7)11-9(3)12/h4-6H,1-3H3,(H,11,12)
Formula Weight 163.0997
Percent Purity >95%
Chemical Name or Material N-(2,6-Dimethylphenyl)-acetamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.