missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Bromophenyl)thiourea, 98%, Thermo Scientific Chemicals

Product Code. 11358225
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11358225 5 g 5g
11342437 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358225 Supplier Thermo Scientific Alfa Aesar Supplier No. L10098.06
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5391-30-0 | Molecular Formula: C7H7BrN2S | Molecular Weight: 231.111 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5391-30-0
Molecular Formula C7H7BrN2S
Molecular Weight (g/mol) 231.111
MDL Number MFCD00041146
InChI Key QIGMVYSPXPXCPN-UHFFFAOYSA-N
Synonym 1-2-bromophenyl thiourea, 1-2-bromophenyl-2-thiourea, 2-bromophenyl thiourea, n-2-bromophenyl thiourea, thiourea, 2-bromophenyl, thiourea,n-2-bromophenyl, thiourea, bromophenyl, amino 2-bromophenyl amino methane-1-thione, o-bromophenylthiourea
PubChem CID 2735618
IUPAC Name (2-bromophenyl)thiourea
SMILES C1=CC=C(C(=C1)NC(=S)N)Br

Specifications

Melting Point 124°C to 126°C
Odor Odorless
Quantity 5 g
UN Number UN2811
Beilstein 2718427
Formula Weight 231.12
Percent Purity 98%
Chemical Name or Material N-(2-Bromophenyl)thiourea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.