missing translation for 'onlineSavingsMsg'
Learn More

N-[(2'-Cyano[1,1'-biphenyl]-4-yl)methyl]-L-valine Methyl Ester Hydrochloride, TRC

Product Code. 30135481
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135481 10 g 10g
30007661 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135481 Supplier Toronto Research Chemicals Supplier No. TRCC96234010G
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 482577-59-3
Molecular Formula C20 H22 N2 O2 . Cl H
Molecular Weight (g/mol) 358.86
Synonym L-Valine, N-[(2'-cyano[1,1'-biphenyl]-4-yl)methyl]-, methyl ester, hydrochloride (1:1), : L-Valine, N-[(2'-cyano[1,1'-biphenyl]-4-yl)methyl]-, methyl ester, monohydrochloride (9CI), N-[(2'-Cyano[1,1'-biphenyl]-4-yl)methyl]-L-valine methyl ester hydrochloride (1:1)
IUPAC Name methyl (2S)-2-[[4-(2-cyanophenyl)phenyl]methylamino]-3-methylbutanoate;hydrochloride
SMILES Cl.COC(=O)[C@@H](NCc1ccc(cc1)c2ccccc2C#N)C(C)C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C20H22N2O2.ClH/c1-14(2)19(20(23)24-3)22-13-15-8-10-16(11-9-15)18-7-5-4-6-17(18)12-21;/h4-11,14,19,22H,13H2,1-3H3;1H/t19-;/m0./s1
Formula Weight 358.1448
Chemical Name or Material N-[(2'-Cyano[1,1'-biphenyl]-4-yl)methyl]-L-valine Methyl Ester Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.