missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Methoxy-5-methylphenyl)thiourea, 99%, Thermo Scientific™

Product Code. 11358615
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358615 1 g 1g
11368615 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358615 Supplier Thermo Scientific Alfa Aesar Supplier No. L12101.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-(2-Methoxy-5-methylphenyl)thiourea is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88686-29-7
Molecular Formula C9H12N2OS
Molecular Weight (g/mol) 196.27
MDL Number MFCD00041174
InChI Key IPKUQBKZOAIUOC-UHFFFAOYSA-N
Synonym 1-2-methoxy-5-methylphenyl-2-thiourea, 1-2-methoxy-5-methylphenyl thiourea, 2-methoxy-5-methylphenyl thiourea, amino 2-methoxy-5-methylphenyl amino methane-1-thione, acmc-20cw9w, 2-methoxy-5-methyl-phenyl-thiourea, n-2-methoxy-5-methylphenyl thiourea, 2-methoxy-5-methylphenylthiourea, thiourea,n-2-methoxy-5-methylphenyl
PubChem CID 736212
IUPAC Name (2-methoxy-5-methylphenyl)thiourea
SMILES COC1=C(NC(N)=S)C=C(C)C=C1

Specifications

Melting Point 127°C to 130°C
Quantity 1 g
UN Number UN2811
Beilstein 2722947
Solubility Information Insoluble in water.
Formula Weight 196.27
Percent Purity 99%
Chemical Name or Material N-(2-Methoxy-5-methylphenyl)thiourea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.