missing translation for 'onlineSavingsMsg'
Learn More

N-(3,5-Dichlorophenyl)benzenesulfonamide, 97%

Product Code. 15476498
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15476498

Brand: Thermo Scientific Alfa Aesar H60031.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 54129-15-6 | C12H9Cl2NO2S | 302.17 g/mol

N-(3,5-dichlorophenyl)benzenesulfonamide (C21) as an inhibitor of Rac activation by Dock5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(3,5-dichlorophenyl)benzenesulfonamide (C21) as an inhibitor of Rac activation by Dock5.

Notes
Reacts with strong oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54129-15-6
Molecular Formula C12H9Cl2NO2S
Molecular Weight (g/mol) 302.17
MDL Number MFCD01212617
InChI Key HMKZVAZQKOKXRZ-UHFFFAOYSA-N
Synonym n-3,5-dichlorophenyl benzenesulfonamide, n-3,5-dichlorophenyl benzensulfonamide, 3,5-dichlorophenyl phenylsulfonyl amine, benzenesulfonamide, n-3,5-dichlorophenyl
PubChem CID 4985978
IUPAC Name N-(3,5-dichlorophenyl)benzenesulfonamide
SMILES ClC1=CC(NS(=O)(=O)C2=CC=CC=C2)=CC(Cl)=C1

Specifications

Color White to Gray
Quantity 250 mg
Formula Weight 302.17
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-(3,5-Dichlorophenyl)benzenesulfonamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.