missing translation for 'onlineSavingsMsg'
Learn More

N-(3,6-Dichloro-2-methylphenyl)-2-(tritylamino)thiazole-5-carboxamide, TRC

Product Code. 30369713
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30369713 5 mg 5mg
30384820 10 mg 10mg
30369542 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30369713 Supplier Toronto Research Chemicals Supplier No. TRCD6305305MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(3,6-Dichloro-2-methylphenyl)-2-(tritylamino)thiazole-5-carboxamide
Analyte or Component Names N-(3,6-Dichloro-2-methylphenyl)-2-(tritylamino)thiazole-5-carboxamide
Shipping Condition Room Temperature
Molecular Formula C30 H23 Cl2 N3 O S
InChI Formula InChI=1S/C30H23Cl2N3OS/c1-20-24(31)17-18-25(32)27(20)34-28(36)26-19-33-29(37-26)35-30(21-11-5-2-6-12-21,22-13-7-3-8-14-22)23-15-9-4-10-16-23/h2-19H,1H3,(H,33,35)(H,34,36)
Solution Type Neat
SMILES Cc1c(Cl)ccc(Cl)c1NC(=O)c2cnc(NC(c3ccccc3)(c4ccccc4)c5ccccc5)s2
IUPAC Name N-(3,6-dichloro-2-methylphenyl)-2-(tritylamino)-1,3-thiazole-5-carboxamide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 544.494
Quantity 5 mg
Formula Weight 543.094
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.