missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Hydroxyphenyl)thiourea, 97%

Product Code. 11392377
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392377 2 g 2g
11302387 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392377 Supplier Thermo Scientific Alfa Aesar Supplier No. L09975.04
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3394-05-6 | C8H8N2O6 | 228.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3394-05-6
MDL Number MFCD00022167

Specifications

Melting Point 175°C to 180°C
Odor Odorless
Molecular Formula C8H8N2O6
Quantity 2 g
Beilstein 2090372
Synonym 4,5-dinitro veratrole, 1,2-dimethoxy-4,5-dinitro-benzene, 4,5-dimethoxy-1,2-dinitrobenzene, 4,5-dinitroveratrole, acmc-1ckho, ghl.pd_mitscher_leg0.933, benzene,1,2-dimethoxy-4,5-dinitro, benzene, 1,2-dimethoxy-4,5-dinitro
InChI Key WFDHPWTYKOAFBJ-UHFFFAOYSA-N
SMILES COC1=C(C=C(C(=C1)[N+](=O)[O-])[N+](=O)[O-])OC
IUPAC Name 1,2-dimethoxy-4,5-dinitrobenzene
Molecular Weight (g/mol) 228.16
PubChem CID 222989
Formula Weight 168.22
Percent Purity 97%
Chemical Name or Material N-(3-Hydroxyphenyl)thiourea
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.