missing translation for 'onlineSavingsMsg'
Learn More

N-[4-(2-Furyl)benzyl]-N-methylamine, ≥95%, Thermo Scientific™

Product Code. 10707105
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10707105 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10707105 Supplier Thermo Scientific Maybridge Supplier No. CC34446DA
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 859850-67-2
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C12H13NO
MDL Number MFCD08060507
Quantity 1 g
Synonym n-4-2-furyl benzyl-n-methylamine, 4-furan-2-yl phenyl methyl methyl amine, benzenemethanamine,4-2-furanyl-n-methyl, 1-4-furan-2-yl phenyl-n-methylmethanamine, 4-2-furyl phenyl methyl methylamine
InChI Key QJHXIMBULPYCDO-UHFFFAOYSA-N
SMILES CNCC1=CC=C(C=C1)C1=CC=CO1
IUPAC Name {[4-(furan-2-yl)phenyl]methyl}(methyl)amine
Molecular Weight (g/mol) 187.24
PubChem CID 7537512
Formula Weight 187.24
Percent Purity 95%
Chemical Name or Material N-[4-(2-Furyl)benzyl]-N-methylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.