missing translation for 'onlineSavingsMsg'
Learn More

N-[4-[(4-Hydroxyphenyl)amino]phenyl]acetamide, TRC

Product Code. 30136211
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136211 1 g 1g
30011148 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136211 Supplier Toronto Research Chemicals Supplier No. TRCH9512301G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93629-82-4
Molecular Formula C14H14N2O2
Molecular Weight (g/mol) 242.27
IUPAC Name N-(4-((4-hydroxyphenyl)amino)phenyl)acetamide
SMILES OC(C=C1)=CC=C1NC2=CC=C(NC(C)=O)C=C2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C14H14N2O2/c1-10(17)15-11-2-4-12(5-3-11)16-13-6-8-14(18)9-7-13/h2-9,16,18H,1H3,(H,15,17)
Formula Weight 242.11
Percent Purity >95%
Chemical Name or Material N-[4-[(4-Hydroxyphenyl)amino]phenyl]acetamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.