missing translation for 'onlineSavingsMsg'
Learn More

N-(4-chloro-2-(2-chlorobenzoyl)phenyl)-2-oxoacetamide, TRC

Product Code. 17913585
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17913585 500 mg 500mg
17943485 50 mg 50mg
17943185 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17913585 Supplier Toronto Research Chemicals Supplier No. TRCC400520500MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C15H9Cl2NO3
Molecular Weight (g/mol) 322.14
IUPAC Name N-(4-chloro-2-(2-chlorobenzoyl)phenyl)-2-oxoacetamide
SMILES O=C(C([H])=O)NC(C=CC(Cl)=C1)=C1C(C2=C(Cl)C=CC=C2)=O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C15H9Cl2NO3/c16-9-5-6-13(18-14(20)8-19)11(7-9)15(21)10-3-1-2-4-12(10)17/h1-8H,(H,18,20)
Formula Weight 321
Chemical Name or Material N-(4-chloro-2-(2-chlorobenzoyl)phenyl)-2-oxoacetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.