missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Chlorobenzyl)-N-methylamine, ≥97%, Thermo Scientific™

Product Code. 10656433
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10656433 1 g Amber Glass Bottle 1g
10424143 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10656433 Supplier Thermo Scientific Maybridge Supplier No. MO00897DA
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 104-11-0
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.625
MDL Number MFCD00018749
InChI Key LMBUJNXYGGNSAH-UHFFFAOYSA-N
Synonym 4-chloro-n-methylbenzylamine, n-methyl-4-chlorobenzylamine, 1-4-chlorophenyl-n-methylmethanamine, n-4-chlorobenzyl-n-methylamine, 4-chloro-benzyl-methyl-amine, 4-chlorophenyl methyl methyl amine, 4-chlorobenzyl methylamine, benzenemethanamine, 4-chloro-n-methyl, 4-chlorobenzylmethylamine, p-chloro-n-methylbenzylamine
PubChem CID 66905
IUPAC Name 1-(4-chlorophenyl)-N-methylmethanamine
SMILES CNCC1=CC=C(C=C1)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 155.63
Percent Purity 97%
Chemical Name or Material N-(4-chlorobenzyl)-N-methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.