missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Cyanophenyl)thiourea, 98%

Product Code. 11388675
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11388675

Brand: Thermo Scientific Alfa Aesar L12219.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3460-55-7 | C8H7N3S | 177.225 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3460-55-7
Molecular Formula C8H7N3S
Molecular Weight (g/mol) 177.225
MDL Number MFCD00041175
InChI Key LFMJROANUIRGBS-UHFFFAOYSA-N
Synonym 4-cyanophenyl thiourea, 1-4-cyanophenyl thiourea, 1-4-cyanophenyl-2-thiourea, n-4-cyanophenyl thiourea, 4-cyanophenyl-thiourea, acmc-1ajd4, thiourea,n-4-cyanophenyl, 4-aminothioxomethyl amino benzenecarbonitrile
PubChem CID 2757973
IUPAC Name (4-cyanophenyl)thiourea
SMILES C1=CC(=CC=C1C#N)NC(=S)N

Specifications

Melting Point ∽205°C (decomposition)
Quantity 1 g
UN Number UN3439
Beilstein 2718720
Formula Weight 177.23
Percent Purity 98%
Chemical Name or Material N-(4-Cyanophenyl)thiourea

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.