missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Ethylbenzyl)-N-methylamine, Thermo Scientific™

Product Code. 10526724
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10526724 5 g Amber Glass Bottle 5g
10196992 250 mg Amber Glass Bottle 250mg
10023642 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10526724 Supplier Thermo Scientific Maybridge Supplier No. MO00891DE
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 568577-84-4
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
InChI Key JCSDSVXBTRWEJS-UHFFFAOYSA-N
Synonym n-methyl-4-ethylbenzylamine, 1-4-ethylphenyl-n-methylmethanamine, n-4-ethylbenzyl-n-methylamine, 4-ethylphenyl methyl methyl amine, benzenemethanamine, 4-ethyl-n-methyl, 1-4-ethylphenyl-n-methyl-methanamine, 4-ethylbenzyl methylamine, 4-ethylphenyl methyl methylamine
PubChem CID 2060575
IUPAC Name 1-(4-ethylphenyl)-N-methylmethanamine
SMILES CCC1=CC=C(C=C1)CNC

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 149.24
Percent Purity 97%
Chemical Name or Material N-(4-Ethylbenzyl)-N-methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.