missing translation for 'onlineSavingsMsg'
Learn More

N-4-Hydroxyphenyl-N'-1,2,3-thiadiazol-5-ylurea, TRC

Product Code. 30402803
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402803 250 mg 250mg
30403817 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30402803 Supplier Toronto Research Chemicals Supplier No. TRCH949385250MG
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65647-48-5
Molecular Formula C9H8N4O2S
Molecular Weight (g/mol) 236.25
Synonym 1,2,3-Thiadiazole Urea Derivatives
IUPAC Name 1-(4-hydroxyphenyl)-3-(thiadiazol-5-yl)urea
SMILES Oc1ccc(NC(=O)Nc2cnns2)cc1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C9H8N4O2S/c14-7-3-1-6(2-4-7)11-9(15)12-8-5-10-13-16-8/h1-5,14H,(H2,11,12,15)
Formula Weight 236.0368
Chemical Name or Material N-4-Hydroxyphenyl-N'-1,2,3-thiadiazol-5-ylurea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.