missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Hydroxyphenyl)thiourea, 98+%

Product Code. 11343226
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11343226 5 g 5g
11333226 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11343226 Supplier Thermo Scientific Alfa Aesar Supplier No. L12537.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1520-27-0 | C7H8N2OS | 168.214 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1520-27-0
Molecular Formula C7H8N2OS
Molecular Weight (g/mol) 168.214
MDL Number MFCD00041186
InChI Key QICKOOCQSYZYQB-UHFFFAOYSA-N
Synonym 1-4-hydroxyphenyl-2-thiourea, 1-4-hydroxyphenyl thiourea, 4-hydroxyphenyl thiourea, p-hydroxyphenylthiourea, usaf b-75, n-4-hydroxyphenyl thiourea, urea, 1-p-hydroxyphenyl-2-thio, thiourea, 4-hydroxyphenyl, 4-hydroxy-phenyl-thiourea, 4-aminothioxomethyl amino phenol
PubChem CID 2759331
IUPAC Name (4-hydroxyphenyl)thiourea
SMILES C1=CC(=CC=C1NC(=S)N)O

Specifications

Melting Point ∽210°C (decomposition)
Odor Odorless
Quantity 5 g
Beilstein 1102429
Formula Weight 168.22
Percent Purity ≥98%
Chemical Name or Material N-(4-Hydroxyphenyl)thiourea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.