missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(4-Methoxybenzyl)-N-methylamine, tech, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
Specifications
Specifications
| Density | 1.0080g/mL |
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Linear Formula | CH3C6H4CH2NCH3 |
| Quantity | 5 g |
| Formula Weight | 151.21 |
| Percent Purity | 97% |
| Chemical Name or Material | N-(4-Methoxybenzyl)-N-methylamine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction