missing translation for 'onlineSavingsMsg'
Learn More

N-[[(4-Methylphenyl)sulfonyl]oxy]ethane -1,2-diamine, ≥95%, Thermo Scientific™

Product Code. 11346935
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11346935 25 g 25g
10394292 1 g 1g
10693793 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11346935 Supplier Thermo Scientific Maybridge Supplier No. SB01735EB
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 175205-36-4
Molecular Formula C9H14N2O3S
Molecular Weight (g/mol) 230.282
MDL Number MFCD00276588
InChI Key RWUMUZOGZPNCIL-UHFFFAOYSA-N
Synonym 2-tosyloxy amino ethanamine, 2-aminoethyl amino 4-methylbenzenesulfonate, benzenesulfonic acid,4-methyl-, 2-aminoethyl azanyl ester, n1-4-methylphenyl sulfonyl oxy ethane-1,2-diamine, 2-aminoethylamino 4-methylbenzenesulfonate, 2-4-methylphenyl sulfonyl-oxy amino ethanamine, n∼1∼-4-methylbenzene-1-sulfonyl oxy ethane-1,2-diamine, n∼1∼-4-methylphenyl sulfonyl oxy ethane-1,2-diamine
PubChem CID 2796248
IUPAC Name (2-aminoethylamino) 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)ONCCN

Specifications

CAS Min % 95.0
Assay Percent Range 95%+
Linear Formula CH3C6H4SO2ONH(CH2)2NH2
Quantity 25 g
Formula Weight 230.29
Percent Purity 95%
Chemical Name or Material N-[[(4-Methylphenyl)sulfonyl]oxy]ethane -1, 2-diamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.