missing translation for 'onlineSavingsMsg'
Learn More

N-(4-tert-butylbenzyl)-N-methylamine, Thermo Scientific™

Product Code. 10566250
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10566250 5 g Amber Glass Bottle 5g
10321530 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10566250 Supplier Thermo Scientific Maybridge Supplier No. MO08598DE
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 65542-26-9
Molecular Formula C12H19N
Molecular Weight (g/mol) 177.291
InChI Key ZSHCHOYJMLEAOX-UHFFFAOYSA-N
Synonym n-4-tert-butylbenzyl-n-methylamine, 4-tert-butylphenyl methyl methyl amine, 1-4-tert-butyl phenyl-n-methylmethanamine, 1-4-tert-butylphenyl-n-methylmethanamine, 4-tert-butylbenzylmethylamine, n-methyl-4-tert-butylbenzylamine, n-methyl-4-tert.butylbenzylamine, n-methyl 4-tert-butylbenzyl amine, 4-tert-butyl phenyl methyl methylamine, benzenemethanamine,4-1,1-dimethylethyl-n-methyl
PubChem CID 2396874
IUPAC Name 1-(4-tert-butylphenyl)-N-methylmethanamine
SMILES CC(C)(C)C1=CC=C(C=C1)CNC

Specifications

Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 177.29
Percent Purity 97%
Chemical Name or Material N-(4-tert-butylbenzyl)-N-methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.