missing translation for 'onlineSavingsMsg'
Learn More

N-(5-Bromopentyl)phthalimide, 97%

Product Code. 11368486
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368486 5 g 5g
11378486 25 g 25g
11388486 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11368486 Supplier Thermo Scientific Alfa Aesar Supplier No. L17543.06
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 954-81-4 | C13H14BrNO2 | 296.164 g/mol

It is a pharmaceutical intermediate and is used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a pharmaceutical intermediate and is used in medicine.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 954-81-4
Molecular Formula C13H14BrNO2
Molecular Weight (g/mol) 296.164
MDL Number MFCD00060522
InChI Key QKVHAKICMNABGB-UHFFFAOYSA-N
Synonym n-5-bromopentyl phthalimide, 2-5-bromopentyl isoindole-1,3-dione, n-5-bromopentyl-phthalimide, 2-5-bromopentyl isoindoline-1,3-dione, 2-5-bromo-pentyl-isoindole-1,3-dione, 2-5-bromopentyl-1h-isoindole-1,3 2h-dione, 2-5-bromopentyl-2,3-dihydro-1h-isoindole-1,3-dione, 5-phthalimidopentyl bromide, 1-bromo-5-phthalimidopentane
PubChem CID 136770
IUPAC Name 2-(5-bromopentyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCCCCBr

Specifications

Melting Point 58°C to 62°C
Odor Odorless
Quantity 5 g
Beilstein 177015
Solubility Information Insoluble in water.
Formula Weight 296.17
Percent Purity 97%
Chemical Name or Material N-(5-Bromopentyl)phthalimide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.