missing translation for 'onlineSavingsMsg'
Learn More

N-(9H-Fluoren-2-ylmethoxycarbonyloxy)succinimide, 98%

Product Code. 10781891 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
Product Code. Quantity Packaging unitSize
10781891 5 g Glass bottle 5g
10606544 1 g Glass bottle 1g
10792495 25 g Glass bottle 25g
3 options
This item is not returnable. View return policy

Product Code. 10781891

Brand: Thermo Scientific Acros 243100050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 82911-69-1 | C19H15NO5 | 337.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82911-69-1
Molecular Formula C19H15NO5
Molecular Weight (g/mol) 337.29
MDL Number MFCD00010733
InChI Key WMSUFWLPZLCIHP-UHFFFAOYSA-N
Synonym fmoc-osu, n-9-fluorenylmethoxycarbonyloxy succinimide, fmoc-onsu, 9-fluorenylmethyl succinimidyl carbonate, fmoc n-hydroxysuccinimide ester, 9h-fluoren-9-yl methyl 2,5-dioxopyrrolidin-1-yl carbonate, 9-fluorenylmethyl n-succinimidyl carbonate, 9-fluorenylmethylsuccinimidyl carbonate, 9-fluorenylmethoxycarbonyl-n-hydroxysuccinimide
PubChem CID 134122
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 9H-fluoren-9-ylmethyl carbonate
SMILES C1CC(=O)N(C1=O)OC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 145.0°C to 153.0°C
Color White
Loss on Drying 0.25% max. (60°C, 2 hrs, 1mm Hg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 337.29
Percent Purity 98%
Physical Form Microfine Powder
Chemical Name or Material N-(9H-Fluoren-2-ylmethoxycarbonyloxy)succinimide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-(9H-Fluoren-2-ylmethoxycarbonyloxy)succinimide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.