missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-D-glucosamine, MedChemExpress

Product Code. 16462019
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16462019 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16462019 Supplier Medchem Express Supplier No. HYA0132
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress N-Acetyl-D-Glucosamine (N-Acetyl-2-amino-2-deoxy-D-glucose) is a monosaccharide derivative of glucose.

N-Acetyl-D-Glucosamine (N-Acetyl-2-amino-2-deoxy-D-glucose) is a monosaccharide derivative of glucose.

Specifications

Chemical Name or Material N-Acetyl-D-glucosamine
Physical Form Powder
CAS 7512-17-6
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C8H15NO6
For Use With (Application) Metabolism-sugar/lipid metabolism
Synonym N-Acetyl-2-amino-2-deoxy-D-glucose
Solubility Information DMSO : 125 mg/mL (565.07 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 100 mg/mL (452.06 mM; Need ultrasonic)
SMILES O=C[C@H](NC(C)=O)[C@H]([C@@H]([C@@H](CO)O)O)O
Molecular Weight (g/mol) 221.21
Formula Weight 221.21
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.