missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl Glufosinate Sodium, TRC

Product Code. 30138252
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138252 500 mg 500mg
17961155 250 mg 250mg
17912005 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138252 Supplier Toronto Research Chemicals Supplier No. TRCA178235500MG
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133659-60-6
Molecular Formula C7 H12 N O5 P . 2 Na
Molecular Weight (g/mol) 267.1272
Synonym Butanoic acid, 2-(acetylamino)-4-(hydroxymethylphosphinyl)-, sodium salt (1:2), Butanoic acid, 2-(acetylamino)-4-(hydroxymethylphosphinyl)-, disodium salt (9CI), 2-(Acetylamino)-4-(hydroxymethylphosphinyl)butanoic acid disodium salt, 2-Acetamido-4-[hydroxy(methyl)phosphoryl]butanoic acid disodium salt, N-Acetyl glufosinate disodium salt, N-Acetyl-DL-glufosinate disodium salt, Glufosinate-N-acetyl disodium
IUPAC Name disodium;2-acetamido-4-[methyl(oxido)phosphoryl]butanoate
SMILES [Na+].[Na+].CC(=O)NC(CCP(=O)(C)[O-])C(=O)[O-]

Specifications

Analyte or Component Names N-Acetyl Glufosinate Sodium
Shipping Condition Room Temperature
InChI Formula InChI=1S/C7H14NO5P.2Na/c1-5(9)8-6(7(10)11)3-4-14(2,12)13;;/h6H,3-4H2,1-2H3,(H,8,9)(H,10,11)(H,12,13);;/q;2*+1/p-2
Solution Type Neat
Industry Type Pharmaceutical
Quantity 500 mg
Formula Weight 267.0248
Percent Purity >95%
Chemical Name or Material N-Acetyl Glufosinate Sodium
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.