missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl Glyphosate-13C2,15N, TRC

Product Code. 17983565
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17983565 1 mg 1mg
17914035 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17983565 Supplier Toronto Research Chemicals Supplier No. TRCA1782471MG
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1346598-31-9
Molecular Formula 13C2 C3 H10 15N O6 P
Molecular Weight (g/mol) 214.09
Synonym Glyphosate-N-acetyl 13C2,15N, Glyphosate-N-acetyl 1,2-13C2 15N
IUPAC Name 2-[acetyl(phosphonomethyl)amino]acetic acid
SMILES CC(=O)[15N]([13CH2][13C](=O)O)CP(=O)(O)O

Specifications

Analyte or Component Names Glyphosate-N-acetyl 13C2,15N
Shipping Condition Room Temperature
InChI Formula InChI=1S/C5H10NO6P/c1-4(7)6(2-5(8)9)3-13(10,11)12/h2-3H2,1H3,(H,8,9)(H2,10,11,12)/i2+1,5+1,6+1
Solution Type Neat
Industry Type Environmental Testing
Quantity 1 mg
Formula Weight 214.0283
Percent Purity >95%
Chemical Name or Material Glyphosate-N-acetyl 13C2,15N
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.