missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-histidine Methyl Ester, TRC

Product Code. 30179475
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179475 250 mg 250mg
30179174 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179475 Supplier Toronto Research Chemicals Supplier No. TRCA178290250MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36097-48-0
Molecular Formula C9 H13 N3 O3
Molecular Weight (g/mol) 211.22
Synonym N-Acetylhistidine Methyl Ester, NSC 232063;
IUPAC Name methyl (2S)-2-acetamido-3-(1H-imidazol-4-yl)propanoate
SMILES COC(=O)[C@H](Cc1c[nH]cn1)NC(=O)C

Specifications

Analyte or Component Names N-Acetyl-L-histidine Methyl Ester
Shipping Condition Room Temperature
InChI Formula InChI=1S/C9H13N3O3/c1-6(13)12-8(9(14)15-2)3-7-4-10-5-11-7/h4-5,8H,3H2,1-2H3,(H,10,11)(H,12,13)/t8-/m0/s1
Solution Type Neat
Industry Type Forensics
Quantity 250 mg
Formula Weight 211.0957
Chemical Name or Material N-Acetyl-L-histidine Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.