missing translation for 'onlineSavingsMsg'
Learn More

N-Acetylornithine, MedChemExpress

Product Code. 16465566
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 16465566

Brand: Medchem Express HY113080

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress N-Acetylornithine is an intermediate in the enzymatic biosynthesis of the amino acid L-arginine from L-glutamate.

N-Acetylornithine is an intermediate in the enzymatic biosynthesis of the amino acid L-arginine from L-glutamate.

Specifications

Chemical Name or Material N-Acetylornithine
Physical Form Powder
CAS 6205-08-9
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C7H14N2O3
For Use With (Application) Metabolism-protein/nucleotide metabolism
Solubility Information H2O : 100 mg/mL (574.05 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES NCCC[C@@H](C(O)=O)NC(C)=O
Molecular Weight (g/mol) 174.2
Formula Weight 174.2
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.