missing translation for 'onlineSavingsMsg'
Learn More

N-Acetylsulfanilyl chloride, 99%

Product Code. 10434565 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10434565 2.5 kg Plastic Bottle 2.5kg
10417740 100 g Glass bottle 100g
10215440 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10434565 Supplier Thermo Scientific Acros Supplier No. 150720025
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 121-60-8 | C8H8ClNO3S | 233.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-60-8
Molecular Formula C8H8ClNO3S
Molecular Weight (g/mol) 233.67
MDL Number MFCD00007442
InChI Key GRDXCFKBQWDAJH-UHFFFAOYSA-N
Synonym n-acetylsulfanilyl chloride, 4-acetamidobenzene-1-sulfonyl chloride, dagenan chloride, 4-acetylamino benzenesulfonyl chloride, 4-acetamidophenylsulfonyl chloride, benzenesulfonyl chloride, 4-acetylamino, acetylsulfanilyl chloride, p-acetamidobenzenesulfonyl chloride, n-acetylsulphanilyl chloride, p-acetaminobenzenesulfonyl chloride
PubChem CID 8481
IUPAC Name 4-acetamidobenzenesulfonyl chloride
SMILES CC(=O)NC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 145.0°C to 148.0°C
Color Beige-Cream to White
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Plastic Bottle
Linear Formula 4-(CH3CONH)C6H4SO2Cl
Quantity 2.5 kg
Beilstein 14, 702
Fieser 01,3
Merck Index 15, 95
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform,ethylene dichloride,,alcohol,ether
Formula Weight 233.67
Percent Purity 99%
Physical Form Granular Crystalline Powder or Crystals
Chemical Name or Material N-Acetylsulfanilyl chloride, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.