missing translation for 'onlineSavingsMsg'
Learn More

N-Alkyl(C12-16)-N-Benzyl-N,N-Dimethylammonium Chloride (Technical Grade), TRC

Product Code. 30008092
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008092 10 g 10g
30011424 5 g 5g
30006432 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008092 Supplier Toronto Research Chemicals Supplier No. TRCA57513810G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68424-85-1
Molecular Formula C17H30ClN
Molecular Weight (g/mol) 283.88
IUPAC Name benzyl-dimethyl-tetradecylazanium;chloride
SMILES CCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1.[Cl-]

Specifications

Quantity 10 g
InChI Formula InChI=1S/C23H42N.ClH/c1-4-5-6-7-8-9-10-11-12-13-14-18-21-24(2,3)22-23-19-16-15-17-20-23;/h15-17,19-20H,4-14,18,21-22H2,1-3H3;1H/q+1;/p-1
Chemical Name or Material N-Alkyl(C12-16)-N-Benzyl-N,N-Dimethylammonium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.