missing translation for 'onlineSavingsMsg'
Learn More

N-alpha-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, 98%

Product Code. 12684997 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12684997 100 mg Glass bottle 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12684997 Supplier Thermo Scientific Acros Supplier No. 439341000
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 95713-52-3 | C9H9FN4O5 | 272.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chiral derivatization reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 95713-52-3
CAS Min % 97.5
CAS Max % 100.0
Melting Point 227.7°C to 229.4°C
Color Yellow to Orange
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C9H9FN4O5
MDL Number MFCD00042049
Quantity 100 mg
Specific Rotation +54.0° to +58.0° (20°C, 589nm) (c=1, acetone)
Synonym marfey's reagent, s-2-5-fluoro-2,4-dinitrophenyl amino propanamide, 2s-2-5-fluoro-2,4-dinitrophenyl amino propanamide, n-2,4-dinitro-5-fluorophenyl-l-alaninamide, 2s-2-5-fluoro-2,4-dinitroanilino propanamide, propanamide, 2-5-fluoro-2,4-dinitrophenyl amino-, 2s, nalpha-2,4-dinitro-5-fluorophenyl-l-alaninamide, n-alpha-2,4-dinitro-5-fluorophenyl-l-alaninamide, fdaa, 1-fd-aa
InChI Key NEPLBHLFDJOJGP-BYPYZUCNSA-N
SMILES C[C@H](NC1=CC(F)=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(N)=O
IUPAC Name (2S)-2-(5-fluoro-2,4-dinitroanilino)propanamide
Molecular Weight (g/mol) 272.19
PubChem CID 5486955
Formula Weight 272.19
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-α-(2, 4-Dinitro-5-fluorophenyl)-L-alaninamide
Show More Show Less
Product Identifier
  • N-alpha-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, 98%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.