missing translation for 'onlineSavingsMsg'
Learn More

N alpha-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, TRC

Product Code. 17970755
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17970755 100 mg 100mg
17992325 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17970755 Supplier Toronto Research Chemicals Supplier No. TRCD479800100MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95713-52-3
Molecular Formula C9 H9 F N4 O5
Molecular Weight (g/mol) 272.19
Synonym (2S)-2-[(5-Fluoro-2,4-dinitrophenyl)amino]propanamide, Propanamide, 2-[(5-fluoro-2,4-dinitrophenyl)amino]-, (S)-, (2S)-2-[(5-Fluoro-2,4-dinitrophenyl)amino]propanamide, 1-Fluoro-2,4-dinitrophenyl-5-L-alanine amide, FDAA, Marfey's reagent, Reagents, Marfey's
IUPAC Name (2S)-2-(5-fluoro-2,4-dinitroanilino)propanamide
SMILES C[C@H](Nc1cc(F)c(cc1[N+](=O)[O-])[N+](=O)[O-])C(=O)N

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H9FN4O5/c1-4(9(11)15)12-6-2-5(10)7(13(16)17)3-8(6)14(18)19/h2-4,12H,1H3,(H2,11,15)/t4-/m0/s1
Formula Weight 272.0557
Chemical Name or Material (2S)-2-(5-Fluoro-2,4-dinitroanilino)propanamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.