missing translation for 'onlineSavingsMsg'
Learn More

N(alpha)-Benzyloxycarbonyl-L-lysine, 98+%

Product Code. 11302626
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11302626

Brand: Thermo Scientific Alfa Aesar L08629.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2212-75-1 | C14H20N2O4 | 280.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2212-75-1
Molecular Formula C14H20N2O4
Molecular Weight (g/mol) 280.32
MDL Number MFCD00038204
InChI Key OJTJKAUNOLVMDX-LBPRGKRZSA-N
Synonym z-lys-oh, nalpha-cbz-l-lysine, n-alpha-cbz-l-lysine, cbz-lys-oh, n-alpha-carbobenzoxy-l-lysine, s-6-amino-2-benzyloxy carbonyl amino hexanoic acid, z-lysine, cbz-l-lysine, n-alpha-benzyloxycarbonyl-l-lysine, lysine, n2-phenylmethoxy carbonyl
PubChem CID 75172
IUPAC Name (2S)-6-amino-2-(phenylmethoxycarbonylamino)hexanoic acid
SMILES NCCCC[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point ∽237°C (decomposition)
Beilstein 2153826
Quantity 1 g
Formula Weight 280.32
Optical Rotation −12° (c=2 in 0.2N HCl)
Percent Purity ≥98%
Chemical Name or Material N(alpha)-Benzyloxycarbonyl-L-lysine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.