missing translation for 'onlineSavingsMsg'
Learn More

N-Benzoyl-2'-deoxycytidine, 98+%

Product Code. 15418927
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15418927 1 g 1g
15438927 250 mg 250mg
15428927 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15418927 Supplier Thermo Scientific Alfa Aesar Supplier No. H52298.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4836-13-9 | C16H17N3O5 | 331.33 g/mol

Reactant in the synthesis of 2', 5'-Dideoxycytidines and Other Derivatives of 2'-Deoxycytidine, N-Protected 2?-Deoxyribonucleosides(useful for oligonucleotide synthesis), 3?-, 5?-and 3?, 5?-di-O-crotonyl 2?-deoxynucleoside derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4836-13-9
Molecular Formula C16H17N3O5
Molecular Weight (g/mol) 331.33
MDL Number MFCD00010115
InChI Key MPSJHJFNKMUKCN-OUCADQQQSA-N
Synonym n4-benzoyl-2'-deoxycytidine, n-benzoyl-2'-deoxy-cytidine, n-benzoyl-2'-deoxycytidine, cytidine, n-benzoyl-2'-deoxy, cytidine,n-benzoyl-2'-deoxy, n-benzoyl-2-deoxy-cytidine, dc-bz, ksc590o7t, n-benzoyl-2/'-deoxycytidine
PubChem CID 9797617
IUPAC Name N-[1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-oxopyrimidin-4-yl]benzamide
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=CC(NC(=O)C2=CC=CC=C2)=NC1=O

Specifications

Packaging Glass bottle
Quantity 1 g
Solubility Information Soluble in methanol (50mg/ml).
Formula Weight 331.33
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material N-Benzoyl-2′-deoxycytidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.