missing translation for 'onlineSavingsMsg'
Learn More

N-Benzoyl-3'-O-(4,4'-dimethoxytrityl)-2'-deoxyadenosine, 97+%

Product Code. 15459725
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15459725 5 g 5g
15469725 250 mg 250mg
15449725 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15459725 Supplier Thermo Scientific Alfa Aesar Supplier No. H52306.06
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 140712-79-4 | C38H35N5O6 | 657.73 g/mol

N-Benzoyl-3'-O-(4,4'-dimethoxytrityl)-2'-deoxyadenosine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzoyl-3′-O-(4,4′-dimethoxytrityl)-2′-deoxyadenosine is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140712-79-4
Molecular Formula C38H35N5O6
Molecular Weight (g/mol) 657.73
MDL Number MFCD04972282
InChI Key LFXBQKFIXWICJR-BLBFAAIZNA-N
Synonym n6-benzoyl-3'-o-4,4'-dimethoxytrityl-2'-deoxyadenosine, adenosine,n-benzoyl-3'-o-bis 4-methoxyphenyl phenylmethyl-2'-deoxy-9ci, n-benzoyl-3'-o-4,4'-dimethoxytrityl-2'-deoxyadenosine, n6-bezoyl-3'-o-4,4'-dimethoxytrityl-2'-deoxyadenosine, n-benzoyl-3'-o-bis 4-methoxyphenyl phenyl methyl-2'-deoxyadenosine, n-9-2r,4s,5r-4-bis 4-methoxyphenyl phenyl methoxy-5-hydroxymethyl tetrahydrofuran-2-yl-9h-purin-6-yl benzamide, n-9-2r,4s,5r-4-bis 4-methoxyphenyl phenyl methoxy-5-hydroxymethyl oxolan-2-yl purin-6-yl benzamide
PubChem CID 15928822
IUPAC Name N-[9-[(2R,4S,5R)-4-[bis(4-methoxyphenyl)-phenylmethoxy]-5-(hydroxymethyl)oxolan-2-yl]purin-6-yl]benzamide
SMILES COC1=CC=C(C=C1)C(O[C@H]1C[C@@H](O[C@@H]1CO)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12)(C1=CC=CC=C1)C1=CC=C(OC)C=C1

Specifications

Quantity 5 g
Formula Weight 657.73
Percent Purity ≥97%
Chemical Name or Material N-Benzoyl-3'-O-(4,4'-dimethoxytrityl)-2'-deoxyadenosine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.